Assay Details
| Gene Symbol: CPSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322270.1 | NP_001309199.1 | 10 - 11 | 1502 | 136 | |
| NM_001322271.1 | NP_001309200.1 | 9 - 10 | 1342 | 136 | ||
| NM_001322272.1 | NP_001309201.1 | 10 - 11 | 1707 | 136 | ||
| NM_017437.2 | NP_059133.1 | 10 - 11 | 1502 | 136 | ||
| GenBank mRNA | AB037788.1 | - | 4 - 5 | 626 | 136 | |
| AK001627.2 | - | 10 - 11 | 1479 | 136 | ||
| AK001912.1 | - | 8 - 9 | 1058 | 136 | ||
| AK023583.1 | - | 7 - 8 | 1079 | 136 | ||
| AL442079.1 | - | 3 - 4 | 401 | 136 | ||
| BC070095.1 | - | 10 - 11 | 1472 | 5267724 | 136 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000568.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14006901.hg.1 | ENST00000298875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006901.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006901.hg.1 | TC14000568.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156635.hg.1 | TC1400008007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156636.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156636.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156636.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156636.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156636.hg.1 | TC1400008007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156637.hg.1 | TC1400008007.hg.1 | Clariom D TC |

CPSF2
Genomic Map