Assay Details
| Gene Symbol: MT1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301267.1 | NP_001288196.1 | 1 - 2 | 98 | 110 | |
| GenBank mRNA | BC020757.1 | - | 1 - 2 | 98 | 4767886 | 110 |
| BM817643.1 | - | 1 - 2 | 98 | 110 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC020757.1,NM_001301267.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000567054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000567054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180010.hg.1 | TC1600010421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010421.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180014.hg.1 | TC1600010421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | ENST00000569500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180015.hg.1 | TC1600010421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | ENST00000568675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180012.hg.1 | TC1600010421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | ENST00000379811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | ENST00000444837.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | NM_001301267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | NM_005950.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180017.hg.1 | TC1600010421.hg.1 | Clariom D TC |

MT1G
Genomic Map