Assay Details
| Gene Symbol: ARHGEF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091281.1 | - | 1 - 2 | 147 | 152 | |
| AK297707.1 | - | 1 - 2 | 173 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK091281.1, AK297707.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |

ARHGEF19
Genomic Map