Assay Details
| Gene Symbol: ARHGEF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153213.3 | NP_694945.2 | 8 - 9 | 1594 | 151 | |
| XM_011540706.2 | XP_011539008.1 | 9 - 10 | 1758 | 151 | ||
| GenBank mRNA | AL137736.1 | - | 3 - 4 | 384 | 151 | |
| BC012982.2 | - | 7 - 8 | 1170 | 151 | ||
| BC040640.1 | - | 8 - 9 | 1594 | 3847806 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002259.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of actin cytoskeleton organization
regulation of Rho protein signal transduction
wound healing
positive regulation of GTPase activity
Wnt signaling pathway, planar cell polarity pathway
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
regulation of actin cytoskeleton organization
regulation of Rho protein signal transduction
wound healing
positive regulation of GTPase activity
Wnt signaling pathway, planar cell polarity pathway
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034473.hg.1 | TC01002259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034473.hg.1 | ENST00000270747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034473.hg.1 | ENST00000375607.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034473.hg.1 | ENST00000478117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034473.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034474.hg.1 | TC01002259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034474.hg.1 | ENST00000270747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034474.hg.1 | ENST00000375607.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034474.hg.1 | ENST00000478117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034474.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196052.hg.1 | TC0100013034.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196051.hg.1 | TC0100013034.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000270747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000441785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000449495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000471928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | ENST00000478210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013034.hg.1 | NM_153213.4 | Refseq |

ARHGEF19
Genomic Map