Assay Details
| Gene Symbol: RPS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC106063.1 | - | 1 - 2 | 102 | 4386992 | 146 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC106063.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009837.hg.1 | ENST00000412454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007965.hg.1 | ENST00000472287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009801.hg.1 | ENST00000482817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000436377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000436377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011821.hg.1 | ENST00000447187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007362.hg.1 | ENST00000461260.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008062.hg.1 | ENST00000495909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010508.hg.1 | ENST00000422817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012365.hg.1 | ENST00000406706.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010760.hg.1 | ENST00000465359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007106.hg.1 | ENST00000445477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013386.hg.1 | ENST00000407538.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011855.hg.1 | ENST00000496294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000436377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000436377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000445817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011821.hg.1 | ENST00000447187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008062.hg.1 | ENST00000495909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000472218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000479802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007693.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013386.hg.1 | ENST00000407538.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000474626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000474973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000476222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000490191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | ENST00000496813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | NM_022551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153186.hg.1 | TC0600007693.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600153175.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153175.hg.1 | ENST00000439602.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153175.hg.1 | TC0600007693.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009837.hg.1 | ENST00000412454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007362.hg.1 | ENST00000461260.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010508.hg.1 | ENST00000422817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011855.hg.1 | ENST00000496294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010760.hg.1 | ENST00000465359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012365.hg.1 | ENST00000406706.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009801.hg.1 | ENST00000482817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009801.hg.1 | NR_077247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007965.hg.1 | ENST00000472287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007106.hg.1 | ENST00000445477.1 | Ensembl |

RPS18
Genomic Map