Assay Details
| Gene Symbol: PRSS53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039503.2 | NP_001034592.1 | 6 - 7 | 884 | 61 | |
| XM_011545816.1 | XP_011544118.1 | 5 - 6 | 2251 | 61 | ||
| XM_011545817.1 | XP_011544119.1 | 5 - 6 | 2251 | 61 | ||
| XM_011545818.2 | XP_011544120.1 | 5 - 6 | 2251 | 61 | ||
| XM_011545819.1 | XP_011544121.1 | 6 - 7 | 2110 | 61 | ||
| XM_011545820.1 | XP_011544122.1 | 6 - 7 | 2110 | 61 | ||
| GenBank mRNA | AJ627035.1 | - | 6 - 7 | 884 | 61 | |
| AK131087.1 | - | 1 - 2 | 3669 | 61 | ||
More Information
Set Membership: |
|

PRSS53
Genomic Map