Assay Details
| Gene Symbol: UBR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541115.2 | XP_011539417.1 | 60 - 61 | 8809 | 83 | |
| XM_017000828.1 | XP_016856317.1 | 58 - 59 | 8674 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

UBR4
Genomic Map