Assay Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143838.2 | NP_001137310.1 | 10 - 11 | 1524 | 138 | |
| GenBank mRNA | BC143689.1 | - | 10 - 11 | 1480 | 9052203 | 138 |
More Information
Set Membership: |
|

SLC13A5
Genomic Map