Assay Details
| Gene Symbol: RBBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006910.4 | NP_008841.2 | 2 - 3 | 1309 | 88 | |
| NM_018703.3 | NP_061173.1 | 2 - 3 | 1309 | 88 | ||
| GenBank mRNA | AB112074.1 | - | 2 - 3 | 313 | 88 | |
| AB112075.1 | - | 2 - 3 | 313 | 88 | ||
| BC063524.1 | - | 2 - 3 | 376 | 88 | ||
| BX648661.1 | - | 2 - 3 | 249 | 88 | ||
More Information
Set Membership: |
|

RBBP6
Genomic Map