Assay Details
| Gene Symbol: NPAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035412.1 | - | 2 | 1786 | 126 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927142 | LOC101927142 | -16.5kb | Sense | Chr. 2 - 100972648 - 100977161 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein L31 | RPL31 | L31 | 6.2kb | Anti-sense | Chr. 2 - 101002229 - 101019693 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013653.hg.1 | ENST00000452364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |

NPAS2
Genomic Map