| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028273.hg.1 |
TC03001550.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028273.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028273.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186208.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300186208.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186208.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300186208.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186208.hg.1 |
TC0300011517.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000295619.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
ENST00000353065.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_001126128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011517.hg.1 |
NM_021935.3 |
Refseq |