Assay Details
| Gene Symbol: PEX26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB242826.1 | - | 1 | 160 | 139 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100192420 | FLJ41941 | -40.3kb | Sense | Chr. 22 - 18029385 - 18037968 on NCBI Build GRCh38 | |
| Downstream | tubulin alpha 8 | TUBA8 | TUBAL2 | 32.0kb | Sense | Chr. 22 - 18110687 - 18131731 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

PEX26
Genomic Map