Assay Details
| Gene Symbol: KAT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199155.1 | NP_001186084.1 | 11 - 12 | 1608 | 142 | |
| NM_001199156.1 | NP_001186085.1 | 10 - 11 | 1281 | 142 | ||
| NM_001199157.1 | NP_001186086.1 | 10 - 11 | 1368 | 142 | ||
| NM_001199158.1 | NP_001186087.1 | 9 - 10 | 1191 | 142 | ||
| NM_007067.4 | NP_008998.1 | 12 - 13 | 1698 | 142 | ||
| XM_011524235.2 | XP_011522537.1 | 6 - 7 | 748 | 142 | ||
| GenBank mRNA | AF074606.1 | - | 12 - 13 | 1524 | 142 | |
| AF140360.1 | - | 12 - 13 | 1621 | 142 | ||
| AF217502.1 | - | 12 - 13 | 1606 | 142 | ||
| AK023890.1 | - | 12 - 13 | 1629 | 142 | ||
| AK122932.1 | - | 12 - 13 | 2064 | 142 | ||
| AK293976.1 | - | 10 - 11 | 1219 | 142 | ||
| AK294014.1 | - | 10 - 11 | 1305 | 142 | ||
| AK294052.1 | - | 9 - 10 | 1117 | 142 | ||
| AK294367.1 | - | 7 - 8 | 807 | 142 | ||
| AK294602.1 | - | 5 - 6 | 720 | 142 | ||
| AK294836.1 | - | 12 - 13 | 1513 | 142 | ||
| AK295914.1 | - | 6 - 7 | 853 | 142 | ||
| AK297212.1 | - | 11 - 12 | 1690 | 142 | ||
| BC032640.2 | - | 12 - 13 | 1608 | 5496142 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

KAT7
Genomic Map