Assay Details
| Gene Symbol: CPAMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015692.2 | NP_056507.2 | 31 - 32 | 4284 | 96 | |
| XM_011527917.1 | XP_011526219.1 | 31 - 32 | 4251 | 96 | ||
| XM_011527918.1 | XP_011526220.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527919.1 | XP_011526221.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527920.2 | XP_011526222.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527921.2 | XP_011526223.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527922.1 | XP_011526224.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527923.1 | XP_011526225.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527924.2 | XP_011526226.1 | 31 - 32 | 4287 | 96 | ||
| XM_011527925.1 | XP_011526227.1 | 15 - 16 | 2129 | 96 | ||
| XM_017026594.1 | XP_016882083.1 | 31 - 32 | 4287 | 96 | ||
| GenBank mRNA | AB033109.1 | - | 31 - 32 | 4109 | 96 | |
| AY101765.1 | - | 31 - 32 | 4128 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001268.hg.1
Set Membership: |
|

CPAMD8
Genomic Map