Assay Details
| Gene Symbol: FLJ32255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104643.1 | 1 - 2 | 2492 | 121 | ||
| GenBank mRNA | AK056817.1 | - | 1 - 2 | 2492 | 121 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074301 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074302 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074294 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902313 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902301 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902310 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902311 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074276 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074272 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 482560 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 479044 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 727388 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P829183 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P829183 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37051 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6388 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16560 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P829183 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P829183 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P829183 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P829181 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01222293 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P70575 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 235292_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 235291_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235292_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235292_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235291_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235291_PM_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235291_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235292_at4 | - | - |
| Human Genome U95 Set | 90162_PM_at | - | - |
| Human Genome U95 Set | 90162_at | - | - |
| Human Genome U133 Set | 235291_PM_s_at | - | - |
| Human Genome U133 Set | 235292_at | - | - |
| Human Genome U133 Set | 235292_PM_at | - | - |
| Human Genome U133 Set | 235291_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1794510 | AK056817.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1745599 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1745599 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794510 | AK056817.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1745599 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794510 | AK056817.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1745599 | - | - |

FLJ32255
Genomic Map