Assay Details
| Gene Symbol: SFTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205854.2 | NP_995326.1 | 1 - 2 | 140 | 170 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001514.hg.1, TC6_cox_hap2000147.hg.1, TC6_dbb_hap3000136.hg.1, TC6_mann_hap4000125.hg.1, TC6_mcf_hap5000124.hg.1, TC6_qbl_hap6000137.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_205854.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002495.hg.1 | TC6_dbb_hap3000136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002495.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002498.hg.1 | TC6_dbb_hap3000136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002498.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002084.hg.1 | TC6_mann_hap4000125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002084.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002535.hg.1 | TC6_qbl_hap6000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002535.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002083.hg.1 | TC6_mann_hap4000125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002083.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002538.hg.1 | TC6_qbl_hap6000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002538.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002231.hg.1 | TC6_mcf_hap5000124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002231.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017150.hg.1 | TC06001514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017150.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017150.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002081.hg.1 | TC6_mann_hap4000125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002081.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002228.hg.1 | TC6_mcf_hap5000124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002228.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002565.hg.1 | TC6_cox_hap2000147.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002565.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002230.hg.1 | TC6_mcf_hap5000124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002230.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017152.hg.1 | TC06001514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017152.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017152.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017153.hg.1 | TC06001514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017153.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017153.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002563.hg.1 | TC6_cox_hap2000147.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002563.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002497.hg.1 | TC6_dbb_hap3000136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002497.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002566.hg.1 | TC6_cox_hap2000147.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002566.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002537.hg.1 | TC6_qbl_hap6000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002537.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011392.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176860.hg.1 | TC0600011392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176864.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176864.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176864.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176864.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176864.hg.1 | TC0600011392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | ENST00000359086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | ENST00000474914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | NM_205854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600176862.hg.1 | TC0600011392.hg.1 | Clariom D TC |

SFTA2
Genomic Map