Assay Details
| Gene Symbol: MRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178817.3 | NP_848932.1 | 3 - 4 | 293 | 148 | |
| NM_206898.1 | NP_996781.1 | 3 - 4 | 293 | 148 | ||
| XM_006724028.3 | XP_006724091.1 | 1 - 2 | 223 | 148 | ||
| XM_017028407.1 | XP_016883896.1 | 1 - 2 | 219 | 148 | ||
| GenBank mRNA | AF454915.1 | - | 3 - 4 | 293 | 148 | |
| AF483549.1 | - | 1 - 2 | 106 | 148 | ||
| AY079152.1 | - | 1 - 2 | 106 | 148 | ||
| BC062721.1 | - | 1 - 2 | 231 | 30327887 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000115.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cAMP biosynthetic process
protein localization to cell surface
brown fat cell differentiation
protein binding
corticotropin hormone receptor binding
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 5 melanocortin receptor binding
type 1 melanocortin receptor binding
positive regulation of cAMP biosynthetic process
protein localization to cell surface
brown fat cell differentiation
protein binding
corticotropin hormone receptor binding
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 5 melanocortin receptor binding
type 1 melanocortin receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | ENST00000399784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | ENST00000399786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000724.hg.1 | TC21000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | ENST00000303645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | ENST00000399784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | ENST00000399786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000728.hg.1 | TC21000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | ENST00000303645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | ENST00000399784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | ENST00000399786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000729.hg.1 | TC21000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | ENST00000399784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | ENST00000399786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000725.hg.1 | TC21000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | TC2100006918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146461.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146459.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146459.hg.1 | TC2100006918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146459.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | TC2100006918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146462.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000303645.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000339944.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000399784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | ENST00000497833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_001285394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_178817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006918.hg.1 | NM_206898.1 | Refseq |

MRAP
Genomic Map