Assay Details
| Gene Symbol: NAP1L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027291.1 | 1 | 1953 | 130 | ||
| GenBank mRNA | AK090915.1 | - | 1 | 1956 | 130 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) binding protein cytoplasmic 1 like 2A | PABPC1L2A | RBM32A | -46.5kb | Sense | Chr. X - 73077338 - 73079512 on NCBI Build GRCh38 |
| Downstream | nucleosome assembly protein 1 like 2 | NAP1L2 | BPX | 85.6kb | Sense | Chr. X - 73212299 - 73214874 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001151.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788135 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17358097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504762 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788171 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 167525 | AF308284.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 167525 | BC036553.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 167525 | NM_001042475.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 973402 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 4427 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3385371 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5102 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5101 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46897 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03717900 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3385371 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3385371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788146 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788146 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788147 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 89076 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 19839 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03717900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788148 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788121 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788135 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358096 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788111 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788143 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788154 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788114 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788146 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358090 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788128 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788166 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788177 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788158 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788133 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788173 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788141 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03717900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015598.hg.1 | TC0X001151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015598.hg.1 | ENST00000373518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015598.hg.1 | NR_027291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010051.hg.1 | ENST00000373518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010051.hg.1 | NR_027291.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168628.hg.1 | ENST00000373518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168628.hg.1 | NR_027291.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168628.hg.1 | ENST00000373518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168628.hg.1 | NR_027291.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168628.hg.1 | TC0X00010051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010051.hg.1 | ENST00000373518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010051.hg.1 | NR_027291.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040069 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040069 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207646 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3298418 | NR_027291.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3298418 | AK090915.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3221888 | NR_027291.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3221888 | AK090915.1 | human_transcriptome |

NAP1L6
Genomic Map