Assay Details
| Gene Symbol: PYM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC009627.1 | - | 1 - 2 | 153 | 104 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000302533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000398213.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000408946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000454792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000547925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000549939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | ENST00000557259.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_001143853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010878.hg.1 | NM_032345.2 | Refseq |

PYM1
Genomic Map