Assay Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099652.1 | NP_001093122.1 | 4 - 5 | 867 | 131 | |
| GenBank mRNA | AK123953.1 | - | 4 - 5 | 763 | 131 | |
| AK293812.1 | - | 2 - 3 | 316 | 131 | ||
| AL834372.1 | - | 5 - 6 | 775 | 131 | ||
| BX647179.1 | - | 2 - 3 | 2388 | 131 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000307.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003234.hg.1 | ENST00000321662.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003234.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003234.hg.1 | TC14000307.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149722.hg.1 | TC1400007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149723.hg.1 | TC1400007168.hg.1 | Clariom D TC |

GPR137C
Genomic Map