Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 461 | LINC00461 |
EyeLinc1, VISC, Visc-1a, Visc-1b, Visc-2 |
Anti-sense | Chr. 5 - 88540779 - 88684803 on NCBI Build GRCh38 | |
| Downstream | myocyte enhancer factor 2C | MEF2C |
C5DELq14.3, DEL5q14.3 |
41.3kb | Anti-sense | Chr. 5 - 88718241 - 88904105 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map