Assay Details
| Gene Symbol: CDC25B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136335.1 | 13 - 14 | 1283 | 158 | ||
| NR_136336.1 | 13 - 14 | 1510 | 158 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CDC25B
Genomic Map