Assay Details
| Gene Symbol: LOC284242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035844.1 | - | 1 | 1221 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 25 member 52 | SLC25A52 | MCART2 | -181.7kb | Anti-sense | Chr. 18 - 31759696 - 31760880 on NCBI Build GRCh38 |
| Downstream | ring finger protein 125 | RNF125 |
TNORS, TRAC-1, TRAC1 |
74.1kb | Sense | Chr. 18 - 32018203 - 32090751 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC284242
Genomic Map