Assay Details
| Gene Symbol: RIOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003831.3 | NP_003822.2 | 7 - 8 | 1431 | 121 | |
| XM_011526243.1 | XP_011524545.1 | 7 - 8 | 1431 | 121 | ||
| GenBank mRNA | AF013591.1 | - | 7 - 8 | 907 | 121 | |
| AK057292.1 | - | 7 - 8 | 1431 | 121 | ||
| AK303938.1 | - | 7 - 8 | 919 | 121 | ||
| AK304571.1 | - | 2 - 3 | 269 | 121 | ||
| BC028360.1 | - | 7 - 8 | 967 | 4828351 | 121 | |
| BC039729.1 | - | 7 - 8 | 946 | 5745005 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000100.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |

RIOK3
Genomic Map