Assay Details
| Gene Symbol: RIOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003831.3 | NP_003822.2 | 9 - 10 | 1791 | 143 | |
| XM_011526242.1 | XP_011524544.1 | 8 - 9 | 1663 | 143 | ||
| XM_011526243.1 | XP_011524545.1 | 9 - 10 | 1648 | 143 | ||
| GenBank mRNA | AF013591.1 | - | 9 - 10 | 1267 | 143 | |
| AK057292.1 | - | 9 - 10 | 1791 | 143 | ||
| AK303938.1 | - | 9 - 10 | 1279 | 143 | ||
| AK304571.1 | - | 4 - 5 | 486 | 143 | ||
| AK310783.1 | - | 3 - 4 | 324 | 143 | ||
| BC028360.1 | - | 9 - 10 | 1327 | 4828351 | 143 | |
| BC039729.1 | - | 9 - 10 | 1306 | 5745005 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000100.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800147198.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800147198.hg.1 | ENST00000581339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800147198.hg.1 | TC1800006889.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800147198.hg.1 | ENST00000581302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800147198.hg.1 | ENST00000581339.1 | Ensembl |

RIOK3
Genomic Map