Assay Details
| Gene Symbol: MIR99AHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027790.3 | 4 - 5 | 474 | 126 | ||
| GenBank mRNA | AK095614.1 | - | 4 - 5 | 474 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

MIR99AHG
Genomic Map