Assay Details
| Gene Symbol: PMS2CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK296451.1 | - | 2 - 3 | 545 | 102 | |
| BC041364.1 | - | 6 - 7 | 625 | 102 | ||
| CR749616.1 | - | 3 - 4 | 209 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296451.1,CR749616.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000348899.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000348899.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000348899.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000348899.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000403974.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | ENST00000431453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013336.hg.1 | NR_002217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |

PMS2CL
Genomic Map