Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250630.3 | XP_005250687.1 | 3 - 4 | 202 | 152 | |
| XM_017012702.1 | XP_016868191.1 | 3 - 4 | 202 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250630.3, XM_017012702.1 transcripts.
Set Membership: |
|

IMMP2L
Genomic Map