Assay Details
| Gene Symbol: RRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301064.1 | NP_001287993.1 | 9 - 10 | 871 | 157 | |
| NM_018427.4 | NP_060897.3 | 10 - 11 | 961 | 157 | ||
| XM_005255376.1 | XP_005255433.1 | 8 - 9 | 736 | 157 | ||
| XM_005255377.4 | XP_005255434.1 | 7 - 8 | 685 | 157 | ||
| XM_011522540.2 | XP_011520842.1 | 5 - 6 | 373 | 157 | ||
| XM_011522541.1 | XP_011520843.1 | 9 - 10 | 800 | 157 | ||
| GenBank mRNA | AF227156.1 | - | 10 - 11 | 927 | 157 | |
| AJ272050.1 | - | 10 - 11 | 881 | 157 | ||
| AK225662.1 | - | 10 - 11 | 937 | 157 | ||
| AK295486.1 | - | 7 - 8 | 554 | 157 | ||
| AK300253.1 | - | 9 - 10 | 780 | 157 | ||
| AK302994.1 | - | 7 - 8 | 608 | 157 | ||
| AK303405.1 | - | 6 - 7 | 454 | 157 | ||
| AK304856.1 | - | 10 - 11 | 827 | 157 | ||
| AK314769.1 | - | 10 - 11 | 875 | 157 | ||
| AK316520.1 | - | 9 - 10 | 785 | 157 | ||
| BC036182.1 | - | 10 - 11 | 876 | 157 | ||
| BC071868.1 | - | 2 - 3 | 196 | 157 | ||
| BC132688.1 | - | 10 - 11 | 878 | 40146710 | 157 | |
| BC132690.1 | - | 10 - 11 | 878 | 40146712 | 157 | |
| DA764092.1 | - | 2 - 3 | 164 | 157 | ||
More Information
Set Membership: |
|

RRN3
Genomic Map