Assay Details
| Gene Symbol: CIITA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720880.3 | XP_006720943.2 | 1 | 238 | 70 | |
| XM_011522484.2 | XP_011520786.1 | 1 | 238 | 70 | ||
| XM_011522485.2 | XP_011520787.1 | 1 | 238 | 70 | ||
| XM_011522486.2 | XP_011520788.1 | 1 | 240 | 70 | ||
| XM_011522487.2 | XP_011520789.1 | 1 | 122 | 70 | ||
| XM_011522489.2 | XP_011520791.1 | 1 | 122 | 70 | ||
| XM_011522491.2 | XP_011520793.1 | 1 | 240 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CIITA
Genomic Map