| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181487.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181487.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181487.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181487.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181487.hg.1 |
TC0500011783.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011783.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181489.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181489.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181489.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181489.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181489.hg.1 |
TC0500011783.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500181490.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181490.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181490.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181490.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181490.hg.1 |
TC0500011783.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500181488.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181488.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181488.hg.1 |
ENST00000504419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181488.hg.1 |
NR_104678.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181488.hg.1 |
TC0500011783.hg.1 |
Clariom D TC |