Assay Details
| Gene Symbol: HERC2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109773.1 | 8 - 9 | 150 | |||
| GenBank mRNA | BX371795.1 | - | 4 - 5 | 585 | 150 | |
| Gene Symbol: LOC102723753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135178.1 | 8 - 9 | 150 | |||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000564730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000566591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | ENST00000568097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011529.hg.1 | NR_109773.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000562687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000564458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007590.hg.1 | ENST00000569697.5 | Ensembl |

HERC2P4
Genomic Map