Assay Details
| Gene Symbol: FBXL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059173.1 | - | 5 - 6 | 519 | 5738074 | 136 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL19| Gene Symbol: FBXL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059173.1 | - | 5 - 6 | 519 | 5738074 | 136 |
Set Membership: |
|