Assay Details
| Gene Symbol: ZDHHC8P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092581.1 | - | 1 - 2 | 192 | 103 | |
| DA485168.1 | - | 1 - 2 | 254 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK092581.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |

ZDHHC8P1
Genomic Map