Assay Details
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011528286.2 | XP_011526588.1 | 2 - 3 | 427 | 146 | |
XM_011528287.2 | XP_011526589.1 | 2 - 3 | 87 | 146 | ||
XM_017027300.1 | XP_016882789.1 | 2 - 3 | 425 | 146 | ||
XM_017027301.1 | XP_016882790.1 | 2 - 3 | 289 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528286.2, XM_011528287.2, XM_017027300.1, XM_017027301.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
