Assay Details
| Gene Symbol: DAW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178821.1 | NP_849143.1 | 9 - 10 | 936 | 151 | |
| XM_011510755.1 | XP_011509057.1 | 10 - 11 | 1545 | 151 | ||
| XM_017003490.1 | XP_016858979.1 | 8 - 9 | 951 | 151 | ||
| GenBank mRNA | AK097409.1 | - | 9 - 10 | 936 | 151 | |
| AK098821.1 | - | 9 - 10 | 884 | 151 | ||
| AK127882.1 | - | 9 - 10 | 1493 | 151 | ||
| AK301861.1 | - | 10 - 11 | 936 | 151 | ||
| BC036377.2 | - | 9 - 10 | 916 | 4824989 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001365.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02021383.hg.1 | TC02001365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021383.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021383.hg.1 | ENST00000373666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021383.hg.1 | ENST00000545118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021383.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200178762.hg.1 | TC0200010982.hg.1 | Clariom D TC |

DAW1
Genomic Map