Assay Details
| Gene Symbol: DAW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011510755.1 | XP_011509057.1 | 1 - 2 | 680 | 91 | |
| GenBank mRNA | AK301861.1 | - | 1 - 2 | 71 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301861.1, XM_011510755.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000309931.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000373666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000440997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000454999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | ENST00000472604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010982.hg.1 | NM_178821.2 | Refseq |

DAW1
Genomic Map