Assay Details
| Gene Symbol: LINC01493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120553.1 | 2 - 3 | 98 | |||
| GenBank mRNA | AI218577.1 | - | 1 - 2 | 98 | ||
| BX099902.1 | - | 2 - 3 | 214 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LINC01493
Genomic Map