Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015257.2 | NP_056072.2 | 2 - 3 | 303 | 152 | |
| XM_011538058.2 | XP_011536360.1 | 2 - 3 | 980 | 152 | ||
| GenBank mRNA | AK294566.1 | - | 2 - 3 | 275 | 152 | |
| BC117436.1 | - | 2 - 3 | 275 | 40125987 | 152 | |
| BC126300.1 | - | 2 - 3 | 275 | 8992016 | 152 | |
| CR749406.1 | - | 2 - 3 | 275 | 152 | ||
More Information
Set Membership: |
|

NEMP1
Genomic Map