Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003677.4 | NP_003668.2 | 7 - 8 | 690 | 104 | |
| GenBank mRNA | AB014731.1 | - | 7 - 8 | 661 | 104 | |
| AF038554.1 | - | 7 - 8 | 686 | 104 | ||
| AF103800.1 | - | 4 - 5 | 409 | 104 | ||
| BC007860.2 | - | 7 - 8 | 620 | 4300744 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000976.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012817.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012817.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012817.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012824.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012824.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012824.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012825.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012825.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012825.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012825.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012819.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012819.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012819.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012823.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012823.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012823.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012821.hg.1 | ENST00000280557.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012821.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012821.hg.1 | TC12000976.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200168422.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168422.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168422.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168422.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168422.hg.1 | TC1200009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168423.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168423.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168423.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168423.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168423.hg.1 | TC1200009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168424.hg.1 | TC1200009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168426.hg.1 | TC1200009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168421.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168421.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168421.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168421.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168421.hg.1 | TC1200009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000280557.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000455982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000537955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | ENST00000539463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009217.hg.1 | NM_003677.4 | Refseq |

DENR
Genomic Map