Assay Details
| Gene Symbol: ZC3H7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017590.5 | NP_060060.3 | 1 - 2 | 273 | 148 | |
| XM_011530016.2 | XP_011528318.1 | 1 - 2 | 254 | 148 | ||
| GenBank mRNA | AF188530.1 | - | 1 - 2 | 101 | 148 | |
| AK225891.1 | - | 1 - 2 | 246 | 148 | ||
| BC098378.1 | - | 1 - 2 | 211 | 148 | ||
| BC152558.1 | - | 1 - 2 | 251 | 8860405 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000335.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF188530.1,BC152558.1,NM_017590.5,XM_011530016.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22006433.hg.1 | ENST00000351589.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006433.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006433.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006433.hg.1 | TC22000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006434.hg.1 | ENST00000351589.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006434.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006434.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006434.hg.1 | TC22000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006429.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006429.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006429.hg.1 | TC22000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006430.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006430.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006430.hg.1 | TC22000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152771.hg.1 | TC2200007467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152771.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152771.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152771.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152771.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152772.hg.1 | TC2200007467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152772.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152772.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152772.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152772.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | TC2200007467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152773.hg.1 | NM_017590.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 179438 | - | - |

ZC3H7B
Genomic Map