Assay Details
| Gene Symbol: ZC3H7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017590.5 | NP_060060.3 | 3 - 4 | 365 | 112 | |
| XM_011530016.2 | XP_011528318.1 | 3 - 4 | 346 | 112 | ||
| GenBank mRNA | AK023849.1 | - | 3 - 4 | 153 | 112 | |
| AK225891.1 | - | 3 - 4 | 338 | 112 | ||
| AK315009.1 | - | 3 - 4 | 307 | 112 | ||
| BC098378.1 | - | 3 - 4 | 303 | 112 | ||
| BC152558.1 | - | 3 - 4 | 343 | 8860405 | 112 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530016.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | TC2200007467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152778.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |

ZC3H7B
Genomic Map