Assay Details
Gene Symbol: OR5L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004739.1 | NP_001004739.1 | 1 | 848 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000440.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216127 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07807014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216095 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216110 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216115 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216109 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216103 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07807004 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216101 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216147 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216128 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07807003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216096 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216129 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07807006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216131 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216122 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452546 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216132 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07807012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216126 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216141 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216104 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28216120 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 250024 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P258235 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P258235 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 15198 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46305 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P32908632 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P258235 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3380702 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P258235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216126 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216103 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216121 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806985 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216130 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216114 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216131 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216110 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216122 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216096 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07807012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216116 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216147 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806975 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07807003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216093 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216127 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216118 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216128 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07807012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216121 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07807003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216114 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216128 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216122 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216126 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806975 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216147 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216093 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216110 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216116 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216130 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216127 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216131 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216096 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216118 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806985 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216103 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28216109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452542 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P258235 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806974 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806971 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806973 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806969 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806976 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P258235 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806973 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806994 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806991 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806974 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806964 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806982 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807045 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807043 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806989 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806997 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807066 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807050 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806947 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807049 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806986 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806979 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807003 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806962 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807051 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807017 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807006 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807073 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807011 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806944 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807000 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806949 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807012 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807055 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806959 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807080 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807071 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807061 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807053 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807047 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807078 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806968 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806976 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807058 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806945 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807069 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806984 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807008 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806999 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807075 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07807014 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452535 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004739_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7939949 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7939948 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1567285_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1567286_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742390_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742390_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11005507.hg.1 | ENST00000378397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005507.hg.1 | NM_001004739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005507.hg.1 | TC11000440.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100153791.hg.1 | ENST00000378397.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100153791.hg.1 | NM_001004739.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100153791.hg.1 | ENST00000378397.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100153791.hg.1 | NM_001004739.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100153791.hg.1 | TC1100007613.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1567286_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567285_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567286_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567285_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001004739_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567285_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567286_at4 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046386 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 187586 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_26671-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 20521 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_26671 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1708583 | NM_001004739.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1708583 | X64990.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2198850 | NM_001004739.1 | human_transcriptome |
