Assay Details
| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099645.1 | NP_001093115.1 | 1 - 2 | 95 | 136 | |
| NR_135259.1 | 1 - 2 | 95 | 136 | |||
| GenBank mRNA | BM451642.1 | - | 1 - 2 | 107 | 136 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001994.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001099645.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007535.hg.1 | ENST00000493291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035654.hg.1 | TC03001994.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035654.hg.1 | ENST00000295830.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035654.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035654.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035646.hg.1 | TC03001994.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035646.hg.1 | ENST00000295830.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035646.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035646.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035655.hg.1 | TC03001994.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035655.hg.1 | ENST00000295830.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035655.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009515.hg.1 | ENST00000508796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035652.hg.1 | TC03001994.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035652.hg.1 | ENST00000295830.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035652.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000478578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013103.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197994.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197994.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197994.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197994.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197994.hg.1 | TC0300013103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009515.hg.1 | ENST00000508796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000295830.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000463836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000466674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000475836.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | ENST00000494771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | NM_001099645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197987.hg.1 | TC0300013103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007535.hg.1 | ENST00000493291.1 | Ensembl |

RPL22L1
Genomic Map