| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035643.hg.1 |
TC03001994.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035643.hg.1 |
ENST00000295830.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035643.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035643.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013103.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000295830.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000463836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000466674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000475836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000478578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
ENST00000494771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
NM_001099645.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197984.hg.1 |
TC0300013103.hg.1 |
Clariom D TC |