Assay Details
| Gene Symbol: PRORSD1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027258.1 | 1 | 92 | |||
| GenBank mRNA | BU607366.1 | - | 1 | 127 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial translational initiation factor 2 | MTIF2 | -14.7kb | Sense | Chr. 2 - 55235583 - 55269311 on NCBI Build GRCh38 | |
| Downstream | coiled-coil domain containing 88A | CCDC88A |
APE, GIRDIN, GIV, GRDN, HkRP1, KIAA1212 |
3.4kb | Sense | Chr. 2 - 55287817 - 55419921 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000322.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 547932 | BC033379.1 | genbank_mrna_human |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P118011 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5081 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5082 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 17740 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48982 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P118011 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P118011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193993 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274565 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274568 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274568 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274565 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21274582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01193993 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193994 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01194005 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15245078 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193993 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193984 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193996 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15245079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193980 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193991 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01194009 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193987 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193973 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193974 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193982 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15245077 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193968 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193966 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01194007 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193988 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193979 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01193990 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35686053 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8042060 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8042061 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8042059 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 237291_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004966.hg.1 | TC02000322.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004966.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004965.hg.1 | TC02000322.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004965.hg.1 | ENST00000295112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004965.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004968.hg.1 | TC02000322.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004968.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004967.hg.1 | TC02000322.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004967.hg.1 | ENST00000295112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004967.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221807.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221807.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221807.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221807.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221807.hg.1 | TC0200016448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221809.hg.1 | TC0200016448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221806.hg.1 | TC0200016448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221808.hg.1 | TC0200016448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000295112.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000563365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | ENST00000579162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016448.hg.1 | NR_027258.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 237291_at | - | - |
| Human Genome U133 Plus 2.0 Array | 237291_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237291_at10 | - | - |
| Human Genome U95 Set | 81735_at | - | - |
| Human Genome U95 Set | 81735_PM_at | - | - |
| Human Genome U133 Set | 237291_at | - | - |
| Human Genome U133 Set | 237291_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015668 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015668 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 200635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2275583 | NR_027258.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2275583 | NR_027258.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2275583 | NR_027258.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2275583 | NR_027258.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1704704 | NR_027258.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2275583 | NR_027258.1 | human_transcriptome |

PRORSD1P
Genomic Map