Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006179.4 | NP_006170.1 | 1 - 2 | 127 | 130 | |
| XM_005258962.3 | XP_005259019.1 | 2 - 3 | 248 | 130 | ||
| XM_006723232.3 | XP_006723295.1 | 2 - 3 | 290 | 130 | ||
| GenBank mRNA | AK297906.1 | - | 2 - 3 | 248 | 130 | |
| BC012421.2 | - | 1 - 2 | 127 | 3865300 | 130 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002718.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297906.1,BC012421.2,NM_006179.4,XM_005258962.3,XM_006723232.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
cell-cell signaling
ganglion mother cell fate determination
long-term memory
sensory organ boundary specification
adult locomotory behavior
epidermis development
mechanoreceptor differentiation
negative regulation of neuron apoptotic process
regulation of neuron differentiation
neuron projection morphogenesis
innervation
taste bud development
neurotrophin p75 receptor binding
protein binding
growth factor activity
transmembrane receptor protein tyrosine kinase signaling pathway
cell-cell signaling
ganglion mother cell fate determination
long-term memory
sensory organ boundary specification
adult locomotory behavior
epidermis development
mechanoreceptor differentiation
negative regulation of neuron apoptotic process
regulation of neuron differentiation
neuron projection morphogenesis
innervation
taste bud development
neurotrophin p75 receptor binding
protein binding
growth factor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19023323.hg.1 | TC19002718.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023323.hg.1 | ENST00000301411.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023323.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011117.hg.1 | ENST00000591175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008515.hg.1 | ENST00000593746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011119.hg.1 | ENST00000591913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023321.hg.1 | TC19002718.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023321.hg.1 | ENST00000301411.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023321.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008512.hg.1 | ENST00000591094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011121.hg.1 | ENST00000596318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | TC1900011998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207049.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008512.hg.1 | ENST00000591094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008515.hg.1 | ENST00000593746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011119.hg.1 | ENST00000591913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000377280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000593309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000596965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000597853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | ENST00000598442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011997.hg.1 | NM_033142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000593537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011998.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011117.hg.1 | ENST00000591175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | TC1900011998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | ENST00000594938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | ENST00000595857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207051.hg.1 | NM_006179.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011121.hg.1 | ENST00000596318.1 | Ensembl |

NTF4
Genomic Map