Assay Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015429.1 | 5 - 6 | 485 | 147 | ||
| NR_024429.1 | 5 - 6 | 581 | 147 | |||
| NR_024431.2 | 7 - 8 | 777 | 147 | |||
| GenBank mRNA | AK098249.1 | - | 7 - 8 | 1222 | 147 | |
| BC102000.1 | - | 4 - 5 | 328 | 147 | ||
| BC102001.1 | - | 4 - 5 | 328 | 147 | ||
| BC105703.1 | - | 4 - 5 | 424 | 147 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 595 | LINC00595 | C10orf101 | -681.0kb | Anti-sense | Chr. 10 - 78267328 - 78280213 on NCBI Build GRCh38 |
| Downstream | zinc finger MIZ-type containing 1 | ZMIZ1 |
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10 |
106.9kb | Anti-sense | Chr. 10 - 79068994 - 79316528 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001448.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018393.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018393.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018393.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018393.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018393.hg.1 | TC10001448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176083.hg.1 | TC1000011152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176081.hg.1 | TC1000011152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176064.hg.1 | TC1000011152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176088.hg.1 | TC1000011152.hg.1 | Clariom D TC |

ZMIZ1-AS1
Genomic Map