Assay Details
| Gene Symbol: TPRKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016058.2 | NP_057142.1 | 2 - 3 | 249 | 110 | |
| XM_017004234.1 | XP_016859723.1 | 2 - 3 | 921 | 110 | ||
| XM_017004235.1 | XP_016859724.1 | 3 - 4 | 703 | 110 | ||
| GenBank mRNA | AF060921.1 | - | 2 - 3 | 188 | 110 | |
| AF151879.1 | - | 2 - 3 | 244 | 110 | ||
| AK291071.1 | - | 2 - 3 | 180 | 110 | ||
| BC029492.1 | - | 2 - 3 | 201 | 5271898 | 110 | |
| DA688967.1 | - | 2 - 3 | 249 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001987.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031663.hg.1 | TC02001987.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031663.hg.1 | ENST00000272424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031663.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031663.hg.1 | ENST00000409716.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031663.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195462.hg.1 | TC0200013075.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000272424.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000318190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000409716.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000462166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000463231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000470012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000484969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000485758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000489476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | ENST00000497464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013075.hg.1 | NM_016058.3 | Refseq |

TPRKB
Genomic Map