Assay Details
| Gene Symbol: GAREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191033.1 | NP_001177962.1 | 5 - 6 | 3207 | 76 | |
| XM_006711951.3 | XP_006712014.1 | 6 - 7 | 1768 | 76 | ||
| GenBank mRNA | AB015349.1 | - | 2 - 3 | 266 | 76 | |
| AK090454.1 | - | 5 - 6 | 3209 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000148.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001696.hg.1 | TC02000148.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001696.hg.1 | ENST00000401533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001696.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001696.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001696.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147792.hg.1 | TC0200007028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147791.hg.1 | TC0200007028.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 210524 | - | - |

GAREM2
Genomic Map